Learn More
Chemglass Life Sciences 1L ChemRxnHub Process System Complete with Glassware, Stand, and Digital Overhead Stirrer
Supplier: Chemglass Life Sciences CG1949X1D
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware. Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.