Learn More
Essendant Low-Speed High Productivity Floor Pads 7300, 18" Diameter, Black, 5/Carton
Supplier: Essendant MMM08276
3M’s most aggressive floor stripping pad, the 3M™ High Productivity Pad 7300, is the best choice for removing four or more coats of finish buildup from hard floors. The stripping pad’s open construction allows stripping solution to circulate through the pad, reducing loading. Continuous nylon filaments, uniformly coated with resin and abrasive particles, provide consistently high performance throughout the life of the pad. For rotary and automatic floor machines. For optimum conformability and pad-to-floor contact, the pad can be backed with a red buffing pad.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.