Learn More
Ted Pella Inc PELCO SEM Storage box and holder for up to 18 of 12.7mm pin mounts
Supplier: Ted Pella Inc 16709
Versatile, cost effective SEM pin mount storage box and holder for pin mounts with a ø3.2mm (1/8") pin. Numbered 1-18 with the spacing designed to accommodate all types of pin mounts with a diameter from 6.4 to 50mm. Can hold 18 of 12.7mm pin mounts, 8 of 18mm pin mounts, 8 of 25mm pin mounts, 2 of 32 or 38 or 50mm pin mounts with space left for the smaller pin mounts. Maximum specimen height between mount surface and closed lid is 16mm (5/8"). Used for FEI/Philips, ZEISS/LEO/Leica, Cambridge CamScan, Tescan, Amray and ETEC SEMs. Box inside dimensions are: 4-5/8" x 2-2/4" x 1-5/16" (11.7 x 7.0 x 3.3cm).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.