Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ 18-Gauge Steel Compartment Sinks
SCI_ED

For washing, rinsing and sanitizing tasks.

Supplier:  Diversified Spaces™ 250480

Catalog No. S37864


Add to Cart

Description

Description

  • Made from 18-gauge T-304 stainless-steel
  • 1.5 in. rounded front and side edges to contain splashes
  • Galvanized 1.62 in. legs with 1 in. adjustable high-impact corrosion resistant feet
  • Accommodate 8 in. on center faucets
  • 9 in. back splash
  • Includes 3.5 in. drain
Specifications

Specifications

Stainless Steel (304, 18 g)
Compartment Sink
Rounded front and side edges
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit