Learn More
Ace Glass, Inc. 15mm Ace-Thred nylon bushing, -110 FETFE O-ring, accepts12.5-14mm OD tubing
Supplier: Ace Glass, Inc. 750606
15mm Ace-Thred nylon bushing connector for joining an Ace-Thred to a reduced-end tube. Bushing accepts 12.5-14mm O.D. tubing. Includes -110 FETFE o-ring.