Learn More
Ace Glass, Inc. 150mL heavy wall filter reactor with three neck head, #50threaded body, 10mm stir shaft and bearing, and 3/8" female NPT 100 micron polyethylene filter adapter.
Supplier: Ace Glass, Inc. 6384115