Learn More
Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
$4955.29
Catalog Number | Mfr. No. | Door Style | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Door Style | Color | Price | Quantity & Availability | ||||
19-822-354
|
Justrite™
8825142 |
Left-hinged, Manual-close | Silver |
Each for $4,955.29
|
|
||||
19-822-353
|
Justrite™
8825172 |
Left-hinged, Manual-close | Light Neutral |
Each for $4,955.29
|
|
||||
19-822-352
|
Justrite™
8825122 |
Left-hinged, Manual-close | Blue |
Each for $4,955.29
|
|
||||
19-822-355
|
Justrite™
8825022 |
Right-hinged, Manual-close | Blue |
Each for $4,955.29
|
|
||||
19-822-357
|
Justrite™
8825042 |
Right-hinged, Manual-close | Silver |
Each for $4,955.29
|
|
||||
19-822-356
|
Justrite™
8825072 |
Right-hinged, Manual-close | Light Neutral |
Each for $4,955.29
|
|
||||
Description
- Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
- Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
- Shelves have the ChemCor thermoplastic coating and provide superior protection against corrosion from drips and off-gassing from harsh chemicals
- Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
Specifications
15 gal. |