Learn More
GUARDIAN EQUIPMENT 15 Gallon Portable Eyewash/Drench Hose Unit
Supplier: GUARDIAN EQUIPMENT G1562
Self-contained eyewash/drench hose unit is ideal for use in low traffic areas, and where a continuous supply of potable water is unavailable for plumbed units. 15 gallon pressurized unit is ideal for floor placement near any hazard, and delivers approximately 10 usable gallons (37.9 liters) of water for 15 minutes, thus complying with the provisions of ANSI Z358.1-2014 for eyewashes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.