Learn More
Chemglass Life Sciences Replacement Washers for 15.2/30 PTFE Plug
Supplier: Chemglass Life Sciences CG45714
Replacement Washers for 15.2/30 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.