Learn More
Chemglass Life Sciences Replacement Nuts for 15.2/30 PTFE Plug
Supplier: Chemglass Life Sciences CG45715
Replacement Nuts for 15.2/30 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.