Learn More
Saf T Pak Inc 95kPa Pressure Bag, 13 x 18in. Printed with Biohazard symbol
Supplier: Saf T Pak Inc 20176
95kPa Pressure Bag, 4-sided sealed bag with tape and pouch, OD: 13 x 18in. ID: 12 x 15.25in. Printed with Biohazard symbol