Learn More
International Crystal 13mm KBr Die Set With 2 SS Anvils & Assorted Paper Inserts for Micro Sample
Supplier: International Crystal 00122477
KBr Die Set, 13mm, Includes: Two stainless steel anvils and assorted paper inserts for micro sample, Material: Stainless steel, Total Height: 2.83 in. (72mm), Plunger Height: 1.36 in. (34.6mm), Base Diameter: 2.0 in. (50.8mm)