Learn More
Foxx Life Sciences UN/DOT Tight Head Containers for Shipping, Transport, Storage, and Waste; 13.5L (3.5 Gallon), HDPE, 70mm Cap, UN 3H1, Bulk Pallet of 128 Units
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 3922001FLS PALLET
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
392-2001-FLS: Ezwaste UN/DOT HDPE Container 13.5L Bulk 128/pallet. EZwaste UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers were made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is available in 13.5L, 20L and 60L. Each comes with a 70S cap. These containers can also be used in other industries including but not limited to sanitation, chemical, agricultural and food. All raw materials used in our containers meet FDA Standards for food contact and UN 3H1 Standards.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.