Learn More
Andwin Scientific 12X75MM PP TUBE 1000/CS
Supplier: Andwin Scientific 60818281
Polypropylene Culture Tube, Without caps. Autoclavable. Size, mm: 12 x 75. Volume: 5mL. 125/bag. Endotoxin-free.