Learn More
Sarstedt Inc 125ML PP URINE BEAKER 500/PK
Supplier: Sarstedt Inc 75.565
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Disposable container 125 ml, with graduation, PP, neutral, length 85 mm, opening diameter 62 mm, 100 pieces in the bag
This item is not returnable.
View return policy