Learn More
Chemglass Life Sciences 125mL Funnel, Addition, Graduated, 14/20 Joint Size, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF055102
125mL Funnel, Addition, Graduated, 14/20 Joint Size, Airfree, Schlenk. Unique design of this addition funnel permits solutions to be prepared using a magnetic Stir Bar. Side arm has a 2mm high vacuum glass T-Bore stopcock having a Stainless Steel constant pressure retention device for evacuation and purging of both the funnel and lower vessel. Lower valve is a 0-4mm Chem-Vac™ Chem-Cap valve. The top outer and lower inner standard taper joints are the same size. The 50mL and 125mL sizes are graduated in 1.0mL subdivisions, while the 250mL has 5.0mL subdivisions.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.