Learn More
Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
Packaged in re-sealable bags making storage and racking simple. Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
Supplier:  Biotix™ 63300078
 View more versions of this product
Catalog No. 12-111-028
Add to Cart
Description
Description
  • Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
  • Achieved through patented technologies including FlexFit and Blade
  • Contain X-Resin giving uniform low retention properties
  • Results in minimal sample retention in tip improving accuracy and precision of results
  • Re-sealable bag
  • Tips must be racked by hand
  • Sterile products undergo E-Beam radiation
Certifications
ISO 9001 , FDA Class I
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Non-Filtered
Bulk
uTip
Product Suggestions
Product Suggestions