Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A O416
Encompass_Preferred
Fabric heating mantle for 12L spherical flask, to 400 degrees C maximum internal operatiing temperature, 2 circuits @ 650W, 115V, CSA
Catalog No. NC9054639
Due to product restrictions, please to purchase or view availability for this product.