Learn More
Andwin Scientific 12 X 75 PP STERILE CAP 1000/CS
Supplier: Andwin Scientific 60818306
The Culture Tubes are constructed with polypropylene. This product is autoclavable, disposable, and sterile. The tubes have a volume capacity of 5 mL, an outer diameter of 12 mm, and length of 75 mm. This is sold in a case of 1000.