Learn More
Waters Corp Polypropylene 12 x 32 mm Screw Neck Vial, with Polyethylene Septumless Cap, 700 µL Volume, 100/pk
Supplier: Waters Corp 186005230
Clean polypropylene-molded vial, preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface. This vial is a cost effective alternative to glass.