Learn More
Electron Microscopy Sciences Spot Plate Polypropylene with 12 Cavities
Supplier: Electron Microscopy Sciences 7157201
Very similar to the white porcelain plate, this PP plate comprises 12 cavities of approximately 1ml capacity and is economically priced. This plate is very high quality, unbreakable as well as autoclavable.