Learn More
Bellco Glass, Inc. 1.1mL Bacteriological/Milk
SDP
Supplier:  Bellco Glass, Inc. 229257000
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bacteriological/Milk pipets are manufactured to meet APHA requirements Specialized graduations allow quick volume identification Color Code Purple 50 Individually Wrapped Case Quantity 250
Catalog No. 50-232-9447