Learn More
Bellco Glass, Inc. 1.1mL Bacteriological/ Milk
Supplier: Bellco Glass, Inc. 229267000
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bacteriological/Milk pipets are manufactured to meet APHA requirements Specialized graduations allow quick volume identification Color Code Purple 50 Individually Wrapped Case Quantity 250