Learn More
Perkin Elmer US LLC 11 mm Pre-Assembled Silver Aluminum Crimp Cap with PTFE/Silicone (Red/White) Septa, 100/pk
Supplier: Perkin Elmer US LLC N9306228
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.