Learn More
Chemglass Life Sciences #11 Internal Thread, Glass Only
Supplier: Chemglass Life Sciences CGB36002
#11 Internal Thread, Glass Only. 16mm Tube OD, Accepts 9-10.5mm OD Tubing. Internal glass thread for use with CG-362, CG-363, CG-364, and CG-366 PTFE bushings. Tubing OD listed is the shank OD of the thread.