Learn More
Azzota Corp 10mm Pathlength Standard Fluorometercuvette 3.5Ml - Pathlength 10mm - Inside Width 10mm Volume 3.5Ml Dimensions 45X12.5X12.5mm; Glass (340Nm-2500Nm)
Supplier: Azzota Corp G204
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
10mm Pathlength Standard FluorometerCuvette 3.5ml - Pathlength 10mm - Inside width 10mm Volume 3.5ml Dimensions 45x12.5x12.5mm; Glass (340nm-2500nm)