Learn More
Waters Corp Flowcell Rebuild Kit, 10 mm
Supplier: Waters Corp WAS081346
The 10mm Flow Cell Rebuild Kit contains the following parts: two lens screws, one entrance lens washer, one entrance lens (curved), ten gaskets, one exit window (flat), one exit lens washer, one cuvette lens screw, one cuvette lens, and one cuvette lens holder.