Learn More
Ace Glass, Inc. 10ML SPECIFIC GRAVITY BOTTLE 10ML SPECIFIC GRAVITY BOTTLE
Supplier: Ace Glass, Inc. 547505
Specific gravity bottle, weld, unadjusted for calibration in the lab. Provided with cap to reduce evaporation losses when using volatile liquids. Stopper is 10/18 and cap 14/20 outer.