Learn More
Chemglass Life Sciences 10mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 14 Thread
Supplier: Chemglass Life Sciences CG186401
10mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 14 Thread. Solid phase peptide synthesis vessel having a medium porosity, fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower stopcock is a “T”-Bore 2mm PTFE to allow nitrogen to be used to upwardly agitate the resin beads while shaking. The O.D. of both lower stems is 8mm. Vessels are available in sizes other than those listed. Please call for details.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.