Learn More
Chemglass Life Sciences 10mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 255mm OAH
Supplier: Chemglass Life Sciences CG171812
10mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 255mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.