Learn More
Thermo Scientific 10ML DIONEX ASE 150/350 CELLUL
Supplier:  Thermo Scientific 088345
Encompass
10mL Dionex ASE 150/350 Cellulose Thimbles, 12mm ID, 50mm height, 1.5mm wall thickness
Catalog No. NC1740089