Learn More
SKC Inc Tedlar Sample Bags, 10 Liters, Dual Stainless Steel Fittings, 10/pk
Supplier: SKC Inc 23110
Tedlar 10-liter Sample Bag with dual stainless steel fittings: the hose/valve for flushing, filling, and sealing the bag, and the septum with a syringe port for removing samples for analysis, pk/10.