Learn More
AirSea USA 10L UN Tinplate Drum Non Lacquer Lined, 1A2/Y1.6/100
Supplier: AirSea USA 520
10L Tinplate drum wihtout lacquer lining that can be placed in code 424 to ship large amounts of UN2814 infectious substances. UN Mark: 1A2/Y1.6/100. 10 L UN Tinplate drum with drop handle and lever locking closure - suitable for shipping dangerous goods in liquid form.