Learn More
Thomas Scientific Cell Strainer, 100 um, Sterile, Yellow, 50/pk
Supplier: Thomas Scientific C4100
Individually Wrapped, with 5 reducing adapters for use with 15mL tubes. Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh.