Learn More
Key Scientific Products Shrings, Shrink Seals, Clear, 100mm Size (Wet)
Supplier: Key Scientific Products BV072
Shrings are wet breathable cellulose bands designed to seal agar plates for long term incubation.