Learn More
Ace Glass, Inc. 100mL powder dispensing funnel vertical type
Supplier: Ace Glass, Inc. 723330
100mL vertical, compact, screw-feed powder dispensing funnel. Features a flexible PTFE screw thread, a #15 Ace-Thred at the top, and a large, 1-1/4 knob. Top side port is 14/20, bottom joint is 24/40. 9-1/2 overall height.
This item is not returnable.
View return policy