Learn More
Chemglass Life Sciences 1000mL Addition Funnel, 29/42 Joint Size, 4mm PTFE Stopcock, 525mm OAH
Supplier: Chemglass Life Sciences CG170207
1000mL Addition Funnel, 29/42 Joint Size, 4mm PTFE Stopcock, 525mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.