Learn More
POLYCARBIN INC Sterile 1000 ul Universal Fit Pipette Tips (Non-Filtered) CS50
Supplier: POLYCARBIN INC PC010404
Polycarbin's pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint. Pipette tips do not contain recycled content, and are universal fit.Rack made from 100% circular economy, recycled contentManufactured in an ISO 9001 compliant facilityCarries a smaller carbon footprintDNase, RNase and Pyrogen freeSterileConfiguration: 10 racks/pack, 5 packs/case (50 racks total)Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, we focused on the tip box, or 'rack,' which is the most dense, carbon-intensive piece of plastic.All tip boxes, of any brand, can be remanufactured and transformed into the next line of lab products using Polycarbin's Alpha Carbin, a mailback service.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.