Learn More
BRUKER BIOSPIN Unracked 7" NMR-tubes with coded closed caps, 5mm, 100/pk
Supplier: BRUKER BIOSPIN Z172600
Set of certified 100 tubes with coded closed caps designed for daily use on NMR or on inner carousel of SampleJet.