Learn More
Ace Glass, Inc. 10 x 20mm porosity D stick GLASSBLOWER
Supplier: Ace Glass, Inc. 1533709
10mm O.D. x 20mm long porous fritted (10-20 micron) glass filter stick, used to fabricate gas disperstion tubes. Hollow at one end. Pack of 6.