Learn More
Cytiva (Formerly Pall Lab) Supor EX ECV membrane filter cartridge, 0.2 m removal rating, 10-inch length, double-o-ring (silicone) with bayonet lock and fin end
Supplier:  Cytiva AB1UECV7PH4
Encompass
High capacity, high flow rate 0.2um sterilizing grade filters for highly efficient filtration of cell harvest material, process intermediates, growth media, buffers and final bulk biological process fluids
Catalog No. NC1921055