Learn More
Ted Pella Inc Unconjugated Gold Sols-Gold Colloids, 10NM, 100 mL
Supplier: Ted Pella Inc 157031
The nanometer sized gold colloids can be characterized as follows: Monodispersed gold nanoparticles supplied in water, having trace amounts of citrate, tannic acid and potassium carbonate. Citrate stabilized with a net negative surface charge. Gold colloid pH's range from above 5 for 5nm gold to less than 9 for the larger sized particles (pH increases with size). Available in a size range from 5 to 250nm. The particles are invaluable tools for light scattering, either Raman or plasmon resonance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.