Learn More
Grainger 10 CC LUER LOCK DISPENSING SYR
Supplier: Grainger 5FVE0
10 cc Luer Lock Dispensing Syringe For Use With Dispensing Needles, Disposable, Reusable. These dispensing syringes feature high density polypropylene construction. Use to dispense adhesive, paste, sealant, grease, or fluids. The syringes can help facilitate careful material placement. Use with compatible dispensing needles and dispensing machines.