Learn More
Thomas Scientific 10,000mL Filter Flask / Bottle Shaped
SDP
Supplier:  Thomas Scientific 1156F84
Encompass
With its thick glass walls and sturdy construction, this 10L bottle/flask is perfect for filtering large volumes of liquid. The thick walls and design enables this flask to withstand vacuum filtration without imploding. Graduated in 1000mL increments on a scale from 2000mL to 8000mL with an accuracy of +/- 5%. Neck size fits a #13 rubber stopper.
Catalog No. NC1617833