Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Octanol, ACS, 99%, Spectrum™ Chemical
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
Specifications
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Residue after Evaporation | 4e-05 |
Percent Purity | 0.99 |
Grade | ACS |
Titratable Acid | 0.0002 meq/g |
CAUTION: For manufacturing or laboratory use only. Not for food use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction