Learn More
Global Resources Inc SS610 SS Container with SS Lid for 1 L Capacity Blender, Stainless Steel
Supplier: Global Resources Inc SS610
Blender 1L offers the quality and consistency you need for blending, mixing or homogenizing a variety of samples: Basic single-speed models feature an on/off switch and a maximum speed of 22,000 rpms.