Learn More
Air Boss Defense FlexAir High Efficiency PAPR System w/Visor, N-60202-WHT
Supplier:  Air Boss Defense N60202WHT
Encompass_Preferred
The FlexAir is a NIOSH-approved, compact, lightweight powered air-purifying respirator (PAPR) providing filtered particulate protection at 99.97% efficiency when used with the AirBoss HE filter. The kit includes 1 FlexAir PAPR Blower White, 1 Charger w/Barrel Plug ‐ Li‐Ion Battery, 1 Belt Assy ‐ 60ʺL x 2ʺW, 1 Airflow Indicator, 1 Breathing Tube Assy ‐ Loose Fitting Head Cover ‐ White, 1 Loose Fitting Visor w/Drawstring ‐ Tychem QC ‐ White, 2 HE Filters, 1 User Instruction ‐ Loose fitting Visor w/Drawstring, 1 User Instruction
Catalog No. NC2744865