Learn More
Air Boss Defense FlexAir ChemBio PAPR System w/SL Hood, N-60207-WHT
Supplier: Air Boss Defense N60207WHT
The FlexAir is a NIOSH-approved, compact, lightweight powered air-purifying respirator (PAPR) providing filtered particulate protection at 99.97% efficiency and removes harmful gases and vapors(HE/AM/CD/CL/FM/HC/HF/MA/OV/SD) when used with the AirBoss M96 filter. 1 FlexAir PAPR Blower White, 1 Charger w/Barrel Plug ‐ Li‐Ion Battery, 1 Belt Assy Decon ‐ 70ʺL x 2ʺW, 1 Airflow Indicator, 1 Breathing Tube Assy ‐ Loose Fitting Head Cover ‐ White, 1 Breathing Tube Cover - Tychem SL, 1 Loose Fitting Hood w/Cape ‐ Tychem SL, 3 M96 Cartridge - NIOSH Approved
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.