Learn More
Perkin Elmer US LLC Sapphire Injector for Demountable ZipTorch with Adapter for Optima 7x00, 1.8 mm I.D.
Supplier: Perkin Elmer US LLC N0782006
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This Demountable Injector is for use with Optima 8x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.