Learn More
Chemglass Life Sciences 1.5in X 1.5in X 1/16in Thick Quartz Plate
SDP
Supplier:  Chemglass Life Sciences CGQ062004
Encompass_Preferred
1.5in X 1.5in X 1/16in Thick Quartz Plate. Commercial quality ground and polished clear fused quartz discs. Edges have a ground finished on all sizes. SIZES OTHER THAN THOSE LISTED ARE AVAILABLE. PLEASE CALL FOR INFORMATION.
Catalog No. CGQ062004